2-Amino-4-methylbenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-5-methylaniline

A214963-100G A214963-10G A214963-1G A214963-25G A214963-5G
77249-194EA 225 USD
77249-194 77249-196 77249-198 77249-200 77249-202
2-Amino-4-methylbenzonitrile 98%
2-Amino-4-methylbenzonitrile

Caution: for Research Use Only

Formula: C₈H₈N₂
MW: 132.17
MDL Number: MFCD00173706
CAS Number: 26830-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR