Nicotinic acid (Niacin) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridine-3-carboxylic acid
, 3-Pyridinecarboxylic acid
, Vitamin B3
, 3-Picolinic acid
, Nicotinic acid
, Niacin
A211899-1000G
A211899-100G
A211899-500G
A211899-10G
A211899-25G
77287-982EA
10
USD
77287-982
77287-980
77287-988
77287-986
77287-984
Nicotinic acid (Niacin) 98%
Nicotinic acid (Niacin)
Caution: for Research Use Only
Formula:
C₆H₅NO₂ MW: 123.11 g/mol |
MDL Number:
MFCD00006391 CAS Number: 59-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...