2'-Deoxyuridine 98%
Supplier: Ambeed
|
Warning
|
A211682-100G
A211682-10G
A211682-25G
A211682-5G
A211682-100MG
77275-174EA
178
USD
77275-174
77275-176
77275-178
77275-180
77748-318
2'-Deoxyuridine 98%
2'-Deoxyuridine
Caution: for Research Use Only
Formula:
C₉H₁₂N₂O₅ |
MDL Number:
MFCD00006527 CAS Number: 951-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...