1-Fluoro-4-ethynylbenzene 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne

A209089-10G A209089-1G A209089-25G A209089-5G
77237-328EA 21 USD
77237-328 77237-330 77237-332 77237-334
1-Fluoro-4-ethynylbenzene 98%
1-Fluoro-4-ethynylbenzene

Caution: for Research Use Only

Formula: C₈H₅F
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR