3'-Aminoacetanilide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(3-Aminophenyl)acetamide, 3-Acetamidoaniline, N-Acetyl-1,3-phenylenediamine, m-Aminoacetanilide
A207281-100G
A207281-25G
77275-024EA
14
USD
77275-024
77303-026
3'-Aminoacetanilide 98%
3'-Aminoacetanilide
Caution: for Research Use Only
Formula:
C₈H₁₀N₂O MW: 150.18 g/mol Boiling Pt: 388 |
MDL Number:
MFCD00025229 CAS Number: 102-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...