2-Aminoquinolin-8-ol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-8-quinolinol

A206158-10G A206158-5G
77260-928EA 265.01 USD
77260-928 77260-930
2-Aminoquinolin-8-ol 95%
2-Aminoquinolin-8-ol

Caution: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD00168931
CAS Number: 70125-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR