2-Acetylbenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetylbenzenecarbonitrile

A195283-100MG A195283-1G A195283-250MG A195283-25G A195283-5G
77260-630EA 6 USD
77260-630 77260-632 77260-634 77260-636 77260-638
2-Acetylbenzonitrile 97%
2-Acetylbenzonitrile

Caution: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00272571
CAS Number: 91054-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR