Guanosine 98%

Supplier: Ambeed
Warning

A176316-100G A176316-10G A176316-25G A176316-500G A176316-1000G
77273-888EA 17 USD
77273-888 77273-890 77273-892 77273-894 77748-152
Guanosine 98%
Guanosine

Caution: for Research Use Only

Formula: C₁₀H₁₃N₅O₅
MDL Number: MFCD00010182
CAS Number: 118-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR