2,3,6-Trifluoroaniline 99%

Supplier: Ambeed
Warning

A171895-1G A171895-5G A171895-25G
77286-904EA 18 USD
77286-904 77286-906 77615-434
2,3,6-Trifluoroaniline 99%
2,3,6-Trifluoroaniline

Caution: for Research Use Only

Formula: C₆H₄F₃N
MW: 147.1
Density: 1.39
MDL Number: MFCD00012369
CAS Number: 67815-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR