1-Methylcyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1-cyclohexanecarboxylic acid, 1-Methylcyclohexane-1-carboxylic acid

A167457-100G A167457-10G A167457-1G A167457-250MG A167457-25G A167457-5G
77236-160EA 561 USD
77236-160 77236-162 77236-164 77236-166 77236-168 77236-170
1-Methylcyclohexanecarboxylic acid 98%
1-Methylcyclohexanecarboxylic acid

Caution: for Research Use Only

Formula: C₈H₁₄O₂
MDL Number: MFCD00001463
CAS Number: 1123-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR