AS601245 98+%

Supplier: Ambeed
Warning

Synonyms: (Z)-2-(benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)imino)-1,2-dihydropyrimidin-4-yl)acetonitrile, (2Z)-2-(3H-1,3-Benzothiazol-2-ylidene)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile, SAPK Inhibitor V, 2-(Benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)amino)pyrimidin-4-yl)acetonitrile, 1,3-Benzothiazol-2-yl-(2-((2-(3-pyridinyl)ethyl)amino)-4-pyrimidinyl)acetonitrile

A163603-10MG A163603-1MG A163603-25MG A163603-50MG A163603-5MG
77235-964EA 99 USD
77235-964 77235-966 77235-968 77235-970 77235-972
AS601245 98+%
AS601245

Caution: for Research Use Only

Formula: C₂₀H₁₆N₆S
MDL Number: MFCD07772196
CAS Number: 345987-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR