(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 97%

Supplier: Ambeed
Warning

A163054-1000G A163054-100G A163054-500G A163054-5G A163054-25G
77273-430EA 165 USD
77273-430 77273-432 77273-434 77273-436 77592-986
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 97%
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid

Caution: for Research Use Only

Formula: C₁₃H₁₅NO₆
MDL Number: MFCD00002801
CAS Number: 1155-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR