3-Amino-3-(3-fluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-3-(3-fluorophenyl)propanoic acid

A162575-1G A162575-25G A162575-5G
77247-684EA 5 USD
77247-684 77247-686 77247-688
3-Amino-3-(3-fluorophenyl)propionic acid 97%
3-Amino-3-(3-fluorophenyl)propionic acid

Caution: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD00764457
CAS Number: 117391-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR