2-Amino-4-(4-bromophenyl)thiazole 98%

Supplier: Ambeed
Warning

A159996-1G A159996-25G A159996-5G
77259-546EA 11 USD
77259-546 77259-548 77259-550
2-Amino-4-(4-bromophenyl)thiazole 98%
2-Amino-4-(4-bromophenyl)thiazole

Caution: for Research Use Only

Formula: C₉H₇BrN₂S
MDL Number: MFCD00051953
CAS Number: 2103-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR