1-Isobutyl-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A150157-1G A150157-250MG
77235-414EA 23 USD
77235-414 77235-416
1-Isobutyl-4-nitrobenzene 97%
1-Isobutyl-4-nitrobenzene

Caution: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00130032
CAS Number: 10342-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR