4-Bromo-N-methyl piperidine 95%

Supplier: Ambeed
Warning

A146858-1G A146858-5G A146858-250MG
77285-960EA 42 USD
77285-960 77549-802 77615-172
4-Bromo-N-methyl piperidine 95%
4-Bromo-N-methyl piperidine

Caution: for Research Use Only

Formula: C₆H₁₂BrN
MDL Number: MFCD00800245
CAS Number: 76444-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR