1-Bromotetradecane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Myristyl bromide
A145420-10G
A145420-100G
A145420-500G
A145420-25G
A145420-1000G
77235-182EA
5.15
USD
77235-182
77235-180
77235-186
77235-184
77235-178
1-Bromotetradecane 98%
1-Bromotetradecane
Caution: for Research Use Only
Formula:
BrH₂C(CH₂)₁₂CH₃ MW: 277.29 g/mol |
MDL Number:
MFCD00000228 CAS Number: 112-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...