2-Chloro-1,4-naphthoquinone 97%

Supplier: Ambeed
Warning

A145307-100G A145307-10G A145307-25G A145307-5G
77272-722EA 25 USD
77272-722 77272-724 77272-726 77272-728
2-Chloro-1,4-naphthoquinone 97%
2-Chloro-1,4-naphthoquinone

Caution: for Research Use Only

Formula: C₁₀H₅ClO₂
MDL Number: MFCD00029187
CAS Number: 1010-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR