4-Bromobenzo[d]thiazol-2-amine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-4-bromobenzo[d]thiazole

A143586-100G A143586-1G A143586-25G A143586-5G
77247-046EA 731 USD
77247-046 77247-048 77247-050 77247-052
4-Bromobenzo[d]thiazol-2-amine 98%
4-Bromobenzo[d]thiazol-2-amine

Caution: for Research Use Only

Formula: C₇H₅BrN₂S
MW: 229.097 g/mol
MDL Number: MFCD04971840
CAS Number: 20358-02-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR