5-Nitrouracil 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nitrouracil, 5-Nitropyrimidine-2,4(1H,3H)-dione, 2,4-Dihydroxy-5-nitropyrimidine
A139617-100G
A139617-10G
A139617-25G
A139617-500G
A139617-5G
77285-676EA
25.75
USD
77285-676
77285-678
77285-680
77285-682
77285-684
5-Nitrouracil 98%
5-Nitrouracil
Caution: for Research Use Only
Formula:
C₄H₃N₃O₄ MW: 157.09 g/mol |
MDL Number:
MFCD00006021 CAS Number: 611-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...