3-(4-Aminophenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminohydrocinnamic acid, 3-(4-Aminophenyl)propanoic acid

A132803-10G A132803-1G A132803-25G A132803-5G
77272-072EA 110 USD
77272-072 77272-074 77272-076 77272-078
3-(4-Aminophenyl)propionic acid 98%
3-(4-Aminophenyl)propionic acid

Caution: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00017118
CAS Number: 2393-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR