AC-55541 98%

Supplier: Ambeed
Warning

Synonyms: (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide

A129091-10MG A129091-1MG A129091-50MG A129091-5MG
77246-450EA 26.78 USD
77246-450 77246-448 77246-446 77246-444
AC-55541 98%
AC-55541

Caution: for Research Use Only

Formula: C₂₅H₂₀BrN₅O₃
MW: 518.3620 g/mol
MDL Number: MFCD00327460
CAS Number: 916170-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR