4'-Acetoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Acetylphenyl acetate, 4-Acetoxyacetophenone, p-Acetylphenyl acetate

A126600-100G A126600-25G A126600-5G A126600-500G
77285-136EA 50 USD
77285-136 77614-988 77614-990 77747-905
4'-Acetoxyacetophenone 98%
4'-Acetoxyacetophenone

Caution: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD00017229
CAS Number: 13031-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR