2-(Hydroxymethyl)indole 97%

Supplier: Ambeed
Warning

Synonyms: (Indole-2-yl)methanol, 2-Indolmethanol, Indole-2-methanol, Indole-2-carbinol, 1H-Indole-2-methanol, (1H-Indol-2-yl)methanol, 2-Indolcarbinol

A122985-10G A122985-1G A122985-250MG A122985-5G
77234-190EA 94.76 USD
77234-190 77234-192 77234-194 77234-196
2-(Hydroxymethyl)indole 97%
2-(Hydroxymethyl)indole

Caution: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00030195
CAS Number: 24621-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR