β-Chloropropiophenone 98%

Supplier: Ambeed
Warning

A121574-100G A121574-10G A121574-25G A121574-5G A121574-500G
77271-530EA 11 USD
77271-530 77271-532 77271-534 77271-536 77614-950
β-Chloropropiophenone 98%
β-Chloropropiophenone

Caution: for Research Use Only

Formula: (C₆H₅)C(O)CH₂CH₂Cl
MDL Number: MFCD00000990
CAS Number: 936-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR