Benzo[b]furan-2-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

A114926-10G A114926-1G A114926-25G A114926-5G A114926-100G
77233-846EA 30 USD
77233-846 77233-848 77233-850 77233-852 77745-850
Benzo[b]furan-2-boronic acid 98%
Benzo[b]furan-2-boronic acid

Caution: for Research Use Only

Formula: C₈H₇BO₃
MDL Number: MFCD00236019
CAS Number: 98437-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR