2,3,4,5,6-Pentafluorobenzyl alcohol 95%
Supplier: Ambeed
|
Warning
|
A112479-10G
A112479-25G
A112479-5G
77245-770EA
21
USD
77245-770
77245-772
77745-830
2,3,4,5,6-Pentafluorobenzyl alcohol 95%
2,3,4,5,6-Pentafluorobenzyl alcohol
Caution: for Research Use Only
Formula:
C₇H₃F₅O |
MDL Number:
MFCD00004602 CAS Number: 440-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...