Cytidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
A107133-1000G
A107133-100G
A107133-25G
A107133-500G
A107133-5G
77270-634EA
135
USD
77270-634
77270-636
77270-638
77270-640
77270-642
77270-644
Cytidine 98%
Cytidine
Caution: for Research Use Only
Formula:
C₉H₁₃N₃O₅ MW: 243.22 g/mol |
MDL Number:
MFCD00006545 CAS Number: 65-46-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...