Tanshinone IIA 97%

Supplier: Ambeed
Warning

A104838-100MG A104838-10MG A104838-1MG A104838-25MG A104838-50MG A104838-5G A104838-5MG A104838-1G A104838-250MG
77283-280EA 46 USD
77283-280 77283-282 77283-284 77283-286 77283-288 77283-290 77283-292 77614-790 77614-792
Tanshinone IIA 97%
Tanshinone IIA

Caution: for Research Use Only

Formula: C₁₉H₁₈O₃
MDL Number: MFCD00238692
CAS Number: 568-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR