2-Amino-5-nitrothiazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine

A102927-1000G A102927-100G A102927-10G A102927-25G A102927-500G
77283-018EA 84 USD
77283-018 77283-020 77283-022 77283-024 77283-026
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole

Caution: for Research Use Only

Formula: C₃H₃N₃O₂S
MW: 145.14 g/mol
Melting Pt: 203
MDL Number: MFCD00005326
CAS Number: 121-66-4
Merck Index: 13,00456

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR