Anti-CNTD1 Rabbit Polyclonal Antibody

Supplier: ProSci
18-118
76763-664EA 435.69 USD
76763-664
Anti-CNTD1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR