1-Adamantaneethanol 98%

Supplier: Ambeed
Warning

A981370-1G A981370-5G A981370-25G
77200-476EA 13 USD
77200-476 77200-478 77618-694
1-Adamantaneethanol 98%
1-Adamantaneethanol

Caution: for Research Use Only

Formula: C₁₂H₂₀O
MDL Number: MFCD00074756
CAS Number: 6240-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR