1,4-Diazabicyclo[2.2.2]octane 98%

Supplier: Ambeed
Danger

A855696-25G A855696-5G
77199-960EA 38.11 USD
77199-960 77199-962 77199-964
1,4-Diazabicyclo[2.2.2]octane 98%
1,4-Diazabicyclo[2.2.2]octane

Caution: for Research Use Only

Formula: C₆H₁₂N₂
MDL Number: MFCD00006689
CAS Number: 280-57-9
UN: 2925
ADR: 4.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR