10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine 98%

Supplier: Ambeed
Warning

A853149-1G A853149-250MG A853149-5G
77171-648EA 45 USD
77171-648 77171-650 77171-652
10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine 98%
10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine

Caution: for Research Use Only

Formula: C₂₁H₁₈BrN
MDL Number: MFCD29919158
CAS Number: 1342892-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR