1-Aminopyrrole 95%

Supplier: Ambeed
Danger

Synonyms: 1H-Pyrrol-1-amine

A839245-100MG A839245-10G A839245-250MG A839245-25G A839245-5G A839245-1G
77199-920EA 5 USD
77199-920 77199-922 77199-924 77199-926 77199-928 77573-666
1-Aminopyrrole 95%
1-Aminopyrrole

Caution: for Research Use Only

Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00059931
CAS Number: 765-39-9
UN: 3267
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR