Apoptosis activator 2 99+%

Supplier: Ambeed
Warning

A804232-10MG A804232-1MG A804232-50MG
77199-632EA 42 USD
77199-632 77199-634 77199-636
Apoptosis activator 2 99+%
Apoptosis activator 2

Caution: for Research Use Only

Formula: C₁₅H₉Cl₂NO₂
MDL Number: MFCD00141556
CAS Number: 79183-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR