2',7'-Difluorofluorescein 98+%

Supplier: Ambeed
Warning

A756917-100MG A756917-25MG A756917-50MG
77171-096EA 303 USD
77171-096 77171-098 77171-100
2',7'-Difluorofluorescein 98+%
2',7'-Difluorofluorescein

Caution: for Research Use Only

Formula: C₂₀H₁₀F₂O₅
MDL Number: MFCD01311849
CAS Number: 195136-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR