10-Methoxy-5H-dibenzo[b,f]azepine 98% (by HPLC)

Supplier: Ambeed
Warning

A727468-100G A727468-1G A727468-25G A727468-500G A727468-5G
77199-046EA 102 USD
77199-046 77199-048 77199-050 77199-052 77199-054
10-Methoxy-5H-dibenzo[b,f]azepine 98% (by HPLC)
10-Methoxy-5H-dibenzo[b,f]azepine

Caution: for Research Use Only

Formula: C₁₅H₁₃NO
MDL Number: MFCD00101079
CAS Number: 4698-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR