1,4-Diiodotetrafluorobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2,4,5-Tetrafluoro-3,6-diiodobenzene
A590430-5G
A590430-1G
77197-884EA
21
USD
77197-884
77197-886
1,4-Diiodotetrafluorobenzene 98%
1,4-Diiodotetrafluorobenzene
Caution: for Research Use Only
Formula:
C₆F₄I₂ MW: 401.87 g/mol |
MDL Number:
MFCD00001055 CAS Number: 392-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...