AGK2 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)acrylamide, 2-Cyano-3-(5-(2,5-dichlorophenyl)furan-2-yl)-N-(quinolin-5-yl)acrylamide

A480363-10MG A480363-1MG A480363-25MG A480363-5MG
77169-124EA 55 USD
77169-124 77169-126 77169-128 77169-130
AGK2 98%
AGK2

Caution: for Research Use Only

Formula: C₂₃H₁₃Cl₂N₃O₂
MW: 434.28 g/mol
MDL Number: MFCD01909444
CAS Number: 304896-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR