Calcium carbonate 98%

Supplier: Ambeed
Warning

A476426-500G A476426-100G
77169-094EA 6 USD
77169-094 77572-498
Calcium carbonate 98%
Calcium carbonate

Caution: for Research Use Only

Formula: CaCO₃
MDL Number: MFCD00010906
CAS Number: 471-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR