3-(4-Chlorophenyl)-1H-pyrazol-5-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine
A465741-100MG
A465741-1G
A465741-250MG
A465741-5G
77169-000EA
9
USD
77169-000
77169-002
77169-004
77169-006
3-(4-Chlorophenyl)-1H-pyrazol-5-amine 98%
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Caution: for Research Use Only
Formula:
C₉H₈ClN₃ MW: 193.64 g/mol |
MDL Number:
MFCD00053046 CAS Number: 78583-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...