3-Acetyl-2,4-dimethylpyrrole 98%

Supplier: Ambeed
Warning

A460827-100MG A460827-250MG
77196-590EA 25 USD
77196-590 77196-592
3-Acetyl-2,4-dimethylpyrrole 98%
3-Acetyl-2,4-dimethylpyrrole

Caution: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00005221
CAS Number: 2386-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR