N-Acetyl-D-galactosamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

A455761-10G A455761-1G A455761-25G A455761-5G A455761-250MG
77196-524EA 76 USD
77196-524 77196-526 77196-528 77196-530 77616-968
N-Acetyl-D-galactosamine 95%
N-Acetyl-D-galactosamine

Caution: for Research Use Only

Formula: C₈H₁₅NO₆
MDL Number: MFCD00065372
CAS Number: 1811-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR