Poly(hexamethylene biguanide) hydrochloride 20% in water

Supplier: Ambeed
Warning

A411573-1000G A411573-100G A411573-25G A411573-500G
77168-442EA 114 USD
77168-442 77168-444 77168-446 77168-448
Poly(hexamethylene biguanide) hydrochloride 20% in water
Poly(hexamethylene biguanide) hydrochloride

Caution: for Research Use Only

Formula: (C₈H₁₇N₅)n·xHCl
MDL Number: MFCD00217750
CAS Number: 32289-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR