(S,S)-(-)-Hydrobenzoin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-1,2-Diphenylethane-1,2-diol

A374830-100G A374830-1G A374830-250MG A374830-25G A374830-5G
77195-522EA 1266 USD
77195-522 77195-524 77195-526 77195-528 77195-530
(S,S)-(-)-Hydrobenzoin 98%
(S,S)-(-)-Hydrobenzoin

Caution: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00064255
CAS Number: 2325-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR