1-Acetylisatin 98%

Supplier: Ambeed
Warning

Synonyms: 1-Acetylindoline-2,3-dione

A349426-1G A349426-5G
77195-196EA 26 USD
77195-196 77195-198
1-Acetylisatin 98%
1-Acetylisatin

Caution: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00158542
CAS Number: 574-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR