4-Aminocyclohexanol hydrochloride 98% (60-70:40-30)

Supplier: Ambeed
Warning

A321832-100G A321832-10G A321832-1G A321832-25G A321832-5G
77167-294EA 221 USD
77167-294 77167-296 77167-298 77167-300 77167-302
4-Aminocyclohexanol hydrochloride 98% (60-70:40-30)
4-Aminocyclohexanol hydrochloride

Caution: for Research Use Only

Formula: C₆H₁₄ClNO
MDL Number: MFCD06410647
CAS Number: 76445-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR