a-Tosylbenzyl isocyanide 98%

Supplier: Ambeed
Danger

Synonyms: (1-Phenyl-1-tosyl)methyl Isocyanide, alpha-(p-Toluenesulfonyl)benzyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide, α-(p-Toluenesulfonyl)benzyl Isocyanide

A313896-1G A313896-250MG A313896-5G A313896-100MG
77167-172EA 93 USD
77167-172 77167-174 77167-176 77892-643
a-Tosylbenzyl isocyanide 98%
a-Tosylbenzyl isocyanide

Caution: for Research Use Only

Formula: C₁₅H₁₃NO₂S
MW: 271.3342 g/mol
MDL Number: MFCD04114776
CAS Number: 36635-66-2
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR