AGI-6780 ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulfonamide, N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide

A254962-10MG A254962-1MG A254962-50MG A254962-5MG A254962-100MG A254962-250MG
77166-062EA 31 USD
77166-062 77166-064 77166-066 77166-068 77683-768 77683-769
AGI-6780 ≥98%, Ambeed.Inc
AGI-6780

Caution: For research use only.

Formula: C₂₁H₁₈F₃N₃O₃S₂
MW: 481.51 g/mol
MDL Number: MFCD26097285
CAS Number: 1432660-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR